missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methylpyrimidine hydrochloride, 97%, Thermo Scientific™

Product Code. 11445906
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5348-51-6
Molecular Formula C5H7ClN2O
Molecular Weight (g/mol) 146.574
MDL Number MFCD00151347
InChI Key FDRYQDWXFZBBAD-UHFFFAOYSA-N
Synonym 4-methylpyrimidin-2-ol hydrochloride, 2-hydroxy-4-methylpyrimidine hydrochloride, 2-hydroxy-4-methylpyrimidine hcl, 6-methylpyrimidin-2 1h-one hydrochloride, 2-hydroxy-4-methylpyrimidine hydro-chloride, 2 1h-pyrimidinone, 4-methyl-, monohydrochloride, 4-methyl-3-hydropyrimidin-2-one, chloride, 2-hydroxy-4-methylpyrimidinehydrochloride, zlchem 746, pubchem22005
PubChem CID 2723648
IUPAC Name 6-methyl-1H-pyrimidin-2-one;hydrochloride
SMILES CC1=CC=NC(=O)N1.Cl

Specifications

Melting Point ∼240°C (decomposition)
Odor Odorless
Quantity 25 g
Beilstein 3691454
Formula Weight 146.58
Percent Purity 97%
Chemical Name or Material 2-Hydroxy-4-methylpyrimidine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'