missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

Product Code. 11411117
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 220239-69-0
Molecular Formula C8H5BrF4
Molecular Weight (g/mol) 257.026
MDL Number MFCD00061174
InChI Key BYRMZRWUEAESEZ-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethyl benzyl bromide, 2-bromomethyl-1-fluoro-4-trifluoromethyl benzene, 2-fluoro-5-trifluoromethylbenzyl bromide, benzene, 2-bromomethyl-1-fluoro-4-trifluoromethyl, acmc-1chyk, ksc496g8r, 2-fluoro-5-trifluoromethylbenzylbromide, 2-fluoro-5-trifluoromethyl benzylbromide
PubChem CID 2737571
IUPAC Name 2-(bromomethyl)-1-fluoro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)CBr)F

Specifications

Refractive Index 1.479
Quantity 1 g
UN Number UN1760
Sensitivity Moisture sensitive
Formula Weight 257.03
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'