missing translation for 'onlineSavingsMsg'
Learn More

2-Dimethylaminoethanethiol hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 10152380
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13242-44-9
Molecular Formula C4H11NS·HCl
Molecular Weight (g/mol) 141.67
MDL Number MFCD00012633
InChI Key NRVFDGZJTPCULU-UHFFFAOYSA-N
Synonym 2-dimethylamino ethanethiol hydrochloride, captamine hydrochloride, meda, captamine hcl, n,n-dimethylcysteamine hydrochloride, dimethylaminoethanethiol hydrochloride, 2-dimethylaminoethanethiol hydrochloride, n-dimethylcysteamine hydrochloride, 2-mercaptoethyl dimethylammonium chloride, unii-m43ax41u87
PubChem CID 25798
IUPAC Name 2-(dimethylamino)ethanethiol;hydrochloride
SMILES CN(C)CCS.Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 154°C to 159°C
Assay Percent Range 94% min. (Iodimetry)
Packaging Glass bottle
Linear Formula (CH3)2NCH2CH2SH·HCl
Quantity 5 g
Solubility Information Solubility in water: soluble in water
Formula Weight 141.67
Percent Purity 95%
Chemical Name or Material 2-Dimethylaminoethanethiol hydrochloride, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Dimethylaminoethanethiol hydrochloride
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.