missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Dimethylaminoethanethiol hydrochloride, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Melting Point | 154°C to 159°C |
Assay Percent Range | 94% min. (Iodimetry) |
Packaging | Glass bottle |
Linear Formula | (CH3)2NCH2CH2SH·HCl |
Quantity | 25 g |
Solubility Information | Solubility in water: soluble in water |
Formula Weight | 141.67 |
Percent Purity | 95% |
Show More |
Safety and Handling
Product Identifier
- 2-Dimethylaminoethanethiol hydrochloride
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction