missing translation for 'onlineSavingsMsg'
Learn More

2-Cyanobenzeneboronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Product Code. 11397339
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214360-48-2
Molecular Formula C13H16BNO2
Molecular Weight (g/mol) 229.086
MDL Number MFCD04038747
InChI Key SKQNWSBNAIOCOC-UHFFFAOYSA-N
Synonym 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-cyanophenylboronic acid pinacol ester, 2-cyanophenylboronic acid, pinacol ester, 2-cyanobenzeneboronic acid pinacol ester, benzonitrile, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 2-2-cyanophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl benzonitrile, pubchem7921, ksc925c3p
PubChem CID 3570229
IUPAC Name 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C#N

Specifications

Melting Point 88°C to 90°C
Quantity 1 g
Formula Weight 229.08
Percent Purity 98%
Chemical Name or Material 2-Cyanobenzeneboronic acid pinacol ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'