Learn More
2-Bromobiphenyl, 98%, Thermo Scientific Chemicals
Description
It is used as an intermediate of organic synthesis and pharmaceutical. It also can react with norborn-2-ene to get 5-phenyl-1,2,3,4,4a,8b-hexahydro-1,4-methanobiphenylene, in synthesis of a novel trispirocyclic hydrocarbon. It is used as a reference compound to investigate extraction efficiency of solid-phase micro extraction fibers, based on methacrylic acid-trimethylolpropanetrimethacrylate copolymers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
It is used as an intermediate of organic synthesis and pharmaceutical. It also can react with norborn-2-ene to get 5-phenyl-1,2,3,4,4a,8b-hexahydro-1,4-methanobiphenylene, in synthesis of a novel trispirocyclic hydrocarbon. It is used as a reference compound to investigate extraction efficiency of solid-phase micro extraction fibers, based on methacrylic acid-trimethylolpropanetrimethacrylate copolymers.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications
Specifications
Melting Point | 0°C to 2°C |
Density | 1.376 |
Boiling Point | 298°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.629 |
Quantity | 5 g |
Beilstein | 1863156 |
Solubility Information | Insoluble in water. |
Formula Weight | 233.12 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only