missing translation for 'onlineSavingsMsg'
Learn More

2'-Bromo-4'-fluoroacetanilide, 98%, Thermo Scientific™

Product Code. 11456187
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1009-22-9
Molecular Formula C8H7BrFNO
Molecular Weight (g/mol) 232.05
MDL Number MFCD00061118
InChI Key JAVSBNOXENOHEI-UHFFFAOYSA-N
PubChem CID 688184
IUPAC Name N-(2-bromo-4-fluorophenyl)acetamide
SMILES CC(=O)NC1=CC=C(F)C=C1Br

Specifications

Melting Point 116°C to 119°C
Quantity 100 g
Beilstein 2803940
Synonym 2'-bromo-4'-fluoroacetanilide, n-2-bromo-4-fluorophenyl acetamide, 2-bromo-4-fluoroacetanilide, n-acetyl 2-bromo-4-fluoroaniline, n-2-bromo-4-fluoro-phenyl acetamide, acetamide, n-2-bromo-4-fluorophenyl, maybridge3_000074, ksc180o6b, javsbnoxenohei-uhfffaoysa, n-acetyl2-bromo-4-fluoroaniline
Formula Weight 232.05
Percent Purity 98%
Chemical Name or Material 2'-Bromo-4'-fluoroacetanilide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.