missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-4,6-difluoroanisole, 97%, Thermo Scientific™

Product Code. 11396609
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202865-59-6
Molecular Formula C7H5BrF2O
Molecular Weight (g/mol) 223.02
MDL Number MFCD00070754
InChI Key MFXWQGHKLXJIIP-UHFFFAOYSA-N
Synonym 2-bromo-4,6-difluoroanisole, 1-bromo-3,5-difluoro-2-methoxy-benzene, buttpark 20\04-77, pubchem2633, acmc-1cq3v, 2,4-difluoro-6-bromoanisole, 2-bromo-4,6-difluoranisol, benzene,1-bromo-3,5-difluoro-2-methoxy
PubChem CID 2736263
IUPAC Name 1-bromo-3,5-difluoro-2-methoxybenzene
SMILES COC1=C(F)C=C(F)C=C1Br

Specifications

Refractive Index 1.505
Quantity 25 g
Formula Weight 223.02
Percent Purity 97%
Chemical Name or Material 2-Bromo-4,6-difluoroanisole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'