missing translation for 'onlineSavingsMsg'
Learn More

2-Arachidonylglycerol, Tocris Bioscience™

Catalog Number 30000769
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Endogenous and non-selective CB agonist; potent GPR55 agonist

Non-selective Cannabinoid Receptor Agonists specifically detects Non-selective Cannabinoid Receptor Agonists in samples. It is validated for .
CAS 53847-30-6
Molecular Formula C23H38O4
MDL Number MFCD01862600
Quantity 10 mg
Synonym 2-arachidonoylglycerol, 2-arachidonylglycerol, 2-ag, 2-arachidonyl-glycerol, 2-arachidonoyl-glycerol, 2-arachidonoyl glycerol, 2-ara-gl, glyceryl 2-arachidonate, 2-monoarachidonoylglycerol, unii-8d239qdw64
InChI Key RCRCTBLIHCHWDZ-DOFZRALJSA-N
SMILES CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCC(=O)OC(CO)CO
IUPAC Name 1,3-dihydroxypropan-2-yl (5Z,8Z,11Z,14Z)-icosa-5,8,11,14-tetraenoate
Molecular Weight (g/mol) 378.55
PubChem CID 5282280
ChEBI CHEBI:52392
Formula Weight 378.55
Percent Purity >98%
Chemical Name or Material 2-Arachidonylglycerol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'