missing translation for 'onlineSavingsMsg'
Learn More

2-aminopyridine-5-boronic acid, 97%, Maybridge

Product Code. 10524994
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10524994

Brand: Thermo Scientific Maybridge MAY00018DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 827614-64-2
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H17BN2O2
MDL Number MFCD05663837
Quantity 1 g
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, 2-aminopyridine-5-boronic acid pinacol ester, 2-amino-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-aminopyridine-5-boronic acid, pinacol ester, 6-aminopyridine-3-boronic acid pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-amine, 2-pyridinamine, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinamine, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-ylamine, 6-aminopyridin-3-ylboronic acid pinacol ester
InChI Key YFTAUNOLAHRUIE-UHFFFAOYSA-N
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(N)N=C1
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine
Molecular Weight (g/mol) 220.08
PubChem CID 2756555
Formula Weight 220.08
Percent Purity 97%
Chemical Name or Material 2-Aminopyridine-5-boronic acid, pinacol ester
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.