Learn More
2-Amino-2-cyanoacetamide, 97+%, Thermo Scientific Chemicals
59.00€ - 241.00€
Chemical Identifiers
CAS | 6719-21-7 |
---|---|
Molecular Formula | C3H5N3O |
Molecular Weight (g/mol) | 99.09 |
MDL Number | MFCD00040532 |
InChI Key | JRWAUKYINYWSTA-UHFFFAOYSA-N |
Synonym | aminocyanoacetamide, 3-nitriloalaninamide, acetamide, 2-amino-2-cyano, 2-aminocyanoacetamide, aminocyanacetamide, 2-amino-cyanoacetamide, 2-amino-2cyanoacetamide, a-amino-a-cyanoacetamide, 2-amino-2-cyanacetamide, 2-amino 2-cyano acetamide |
PubChem CID | 98341 |
IUPAC Name | 2-amino-2-cyanoacetamide |
SMILES | C(#N)C(C(=O)N)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10168152
|
Thermo Scientific Acros
382760050 |
5 g | Glass bottle |
59.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10270103
|
Thermo Scientific Acros
382760250 |
25 g | Glass bottle |
241.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6719-21-7 | |
99.09 | |
JRWAUKYINYWSTA-UHFFFAOYSA-N | |
98341 | |
C(#N)C(C(=O)N)N |
C3H5N3O | |
MFCD00040532 | |
aminocyanoacetamide, 3-nitriloalaninamide, acetamide, 2-amino-2-cyano, 2-aminocyanoacetamide, aminocyanacetamide, 2-amino-cyanoacetamide, 2-amino-2cyanoacetamide, a-amino-a-cyanoacetamide, 2-amino-2-cyanacetamide, 2-amino 2-cyano acetamide | |
2-amino-2-cyanoacetamide |
Specifications
6719-21-7 | |
100.0 | |
Beige to Tan | |
97+% | |
NCCH(NH2)CONH2 | |
aminocyanoacetamide, 3-nitriloalaninamide, acetamide, 2-amino-2-cyano, 2-aminocyanoacetamide, aminocyanacetamide, 2-amino-cyanoacetamide, 2-amino-2cyanoacetamide, a-amino-a-cyanoacetamide, 2-amino-2-cyanacetamide, 2-amino 2-cyano acetamide | |
JRWAUKYINYWSTA-UHFFFAOYSA-N | |
2-amino-2-cyanoacetamide | |
98341 | |
97+% | |
2-Amino-2-cyanoacetamide |
97.0 | |
120°C to 124°C | |
Authentic | |
C3H5N3O | |
MFCD00040532 | |
Solubility in water: soluble. Other solubilities: soluble in ether and methanol | |
C(#N)C(C(=O)N)N | |
99.09 | |
99.09 | |
Powder |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 229-771-3
- 2-Amino-2-cyanoacetamide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only