missing translation for 'onlineSavingsMsg'
Learn More

2-Acetoxyisobutyryl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11374384
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11374384

Brand: Thermo Scientific Alfa Aesar L00957.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40635-66-3
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.585
MDL Number MFCD00000708
InChI Key RBTCRFLJLUNCLL-UHFFFAOYSA-N
Synonym 2-acetoxyisobutyryl chloride, 1-chlorocarbonyl-1-methylethyl acetate, 2-acetoxy-2-methylpropionyl chloride, propanoyl chloride, 2-acetyloxy-2-methyl, 2-acetoxyisobutyroyl chloride, 2-chloro-1,1-dimethyl-2-oxoethyl acetate, acmc-1an8z, 2-acetoxyisobutyrylchloride, acetoxyisobutyric acid chloride, acetoxy-iso-butyrylchloride
PubChem CID 123496
IUPAC Name (1-chloro-2-methyl-1-oxopropan-2-yl) acetate
SMILES CC(=O)OC(C)(C)C(=O)Cl

Specifications

Density 1.15
Boiling Point 55°C to 56°C (6 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.429
Quantity 10 g
UN Number UN3265
Beilstein 507772
Sensitivity Moisture sensitive
Formula Weight 164.59
Percent Purity 98%
Chemical Name or Material 2-Acetoxyisobutyryl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.