missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethyl-3(2H)-furanone, 94%, Thermo Scientific Chemicals

Product Code. 11464823
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14400-67-0
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00052571
InChI Key ASOSVCXGWPDUGN-UHFFFAOYSA-N
Synonym 2,5-dimethyl-3 2h-furanone, 2,5-dimethylfuran-3 2h-one, 2,5-dimethyl-2,3-dihydrofuran-3-one, 3 2h-furanone, 2,5-dimethyl, 2,5-dimethyl-3 2h furanone, 2,5-dimethyl-2h-furan-3-one, 2,5-dimethyl-3-2h-furanone, 2h-furan-3-one, 2,5-dimethyl, mango furanone
PubChem CID 85730
IUPAC Name 2,5-dimethylfuran-3-one
SMILES CC1C(=O)C=C(O1)C

Specifications

Density 1.06
Boiling Point 259°C to 261°C
Flash Point 68°C (154°F)
Odor Sweet
Quantity 5 g
Formula Weight 112.13
Percent Purity 94%
Chemical Name or Material 2,5-Dimethyl-3 (2H)-furanone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'