missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Nitrophenyl)ethanol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
It is a phosphate protecting reagent and O6 of deoxyguanosine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a phosphate protecting reagent and O6 of deoxyguanosine.
Solubility
Slightly soluble in water.
Notes
Incompatible with oxidizing agents. Keep at room temperature.
It is a phosphate protecting reagent and O6 of deoxyguanosine.
Solubility
Slightly soluble in water.
Notes
Incompatible with oxidizing agents. Keep at room temperature.
Specifications
Specifications
Melting Point | 59°C to 62°C |
Boiling Point | 177°C (16 mmHg) |
Quantity | 1 g |
Beilstein | 1866148 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 167.16 |
Percent Purity | 98% |
Chemical Name or Material | 2-(4-Nitrophenyl)ethanol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'