missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triisopropylbenzenesulfonyl chloride, 98%

Product Code. 11460333
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11460333

Brand: Thermo Scientific Alfa Aesar A11458.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6553-96-4 | C15H23ClO2S | 302.857 g/mol

2,4,6-Triisopropylbenzenesulfonyl chloride is used for the synthesis of glycerophospholipids as a condensing agent and in the analysis of phosphonolipids in egg yolk by HPLC/MS technique. It acts as a coupling reagent for the synthesis of oligonucleotides and as sulfonates of pyrimidine nucleosides. It also serves as a condensing agent in the synthesis of hydrogen-phosphonate diesters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Triisopropylbenzenesulfonyl chloride is used for the synthesis of glycerophospholipids as a condensing agent and in the analysis of phosphonolipids in egg yolk by HPLC/MS technique. It acts as a coupling reagent for the synthesis of oligonucleotides and as sulfonates of pyrimidine nucleosides. It also serves as a condensing agent in the synthesis of hydrogen-phosphonate diesters.

Solubility
Soluble in chloroform.

Notes
Incompatible with strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6553-96-4
Molecular Formula C15H23ClO2S
Molecular Weight (g/mol) 302.857
MDL Number MFCD00007433
InChI Key JAPYIBBSTJFDAK-UHFFFAOYSA-N
Synonym 2,4,6-triisopropylbenzenesulfonyl chloride, 2,4,6-triisopropylbenzene-1-sulfonyl chloride, trisyl chloride, 2,4,6-triisopropylbenzenesulphonyl chloride, benzenesulfonyl chloride, 2,4,6-tris 1-methylethyl, 2,4,6-triisopropylbenzenesulfonylchloride, 2,4,6-triisopropyl-benzenesulfonyl chloride, tpscl 2,4,6-triisopropyl benzenesulfonyl chloride, 2,4,6-triisopropylphenylsulfonyl chloride, 2,4,6-triisopropyl benzenesulfonyl chloride
PubChem CID 81042
IUPAC Name 2,4,6-tri(propan-2-yl)benzenesulfonyl chloride
SMILES CC(C)C1=CC(=C(C(=C1)C(C)C)S(=O)(=O)Cl)C(C)C

Specifications

Melting Point 94°C to 97°C
Quantity 100 g
UN Number UN3261
Beilstein 1218575
Sensitivity Moisture sensitive
Solubility Information Soluble in chloroform.
Formula Weight 302.87
Percent Purity 98%
Chemical Name or Material 2,4,6-Triisopropylbenzenesulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.