missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloromaleic anhydride, 97%

Product Code. 15535866 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15535866

Brand: Thermo Scientific Acros 113440250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CAS: 1122-17-4 | C4Cl2O3 | 166.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-17-4
Molecular Formula C4Cl2O3
Molecular Weight (g/mol) 166.95
InChI Key AGULWIQIYWWFBJ-UHFFFAOYSA-N
Synonym dichloromaleic anhydride, 2,3-dichloromaleic anhydride, 2,5-furandione, 3,4-dichloro, maleic anhydride, dichloro, dichloromaleic acid anhydride, dichloromaleicanhydride, dichlorofuran-2,5-dione, 2,3-dichloromaleic acid anhydride, dichloro-2,5-dihydrofuran-2,5-dione, 2, 3,4-dichloro
PubChem CID 70728
IUPAC Name 3,4-dichlorofuran-2,5-dione
SMILES C1(=C(C(=O)OC1=O)Cl)Cl

Specifications

Melting Point 115.0°C to 117.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 25 g
Beilstein 17,434
Fieser 04,135; 05,195
Formula Weight 166.95
Percent Purity 97%
Chemical Name or Material 2,3-Dichloromaleic anhydride
compliance-icons
Product Identifier
  • 2,3-Dichloromaleic anhydride, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.