missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorophenoxyacetic acid, 98+%, Thermo Scientific™

Product Code. 11409747
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14892-14-9
Molecular Formula C8H3F5O3
Molecular Weight (g/mol) 242.101
MDL Number MFCD00004297
InChI Key SMXPFEBIAASLOR-UHFFFAOYSA-N
Synonym pentafluorophenoxyacetic acid, pentafluorophenoxy acetic acid, 2,3,4,5,6-pentafluorophenoxyacetic acid, pentafluorophenyloxyacetic acid, acetic acid, pentafluorophenoxy, 2,3,4,5,6-pentafluorophenoxy acetic acid, 2-2,3,4,5,6-pentafluorophenoxy acetic acid, pentafluorophenyloxy-acetic acid
PubChem CID 84680
IUPAC Name 2-(2,3,4,5,6-pentafluorophenoxy)acetic acid
SMILES C(C(=O)O)OC1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Melting Point 108°C to 110°C
Quantity 1 g
Beilstein 1888139
Formula Weight 242.1
Percent Purity ≥98%
Chemical Name or Material 2,3,4,5,6-Pentafluorophenoxyacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'