missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Hydroxyethyl)pyridine, 98%, Thermo Scientific Chemicals

Product Code. 11485607
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

2-(2-Hydroxyethyl)pyridine is a metabolite of the anti-vertigo drug Betahistine. It acts as a reagent for protection of the 5'-phosphate of nucleotides as 2-pyridylethyl (Pet) esters. It is used for carboxyl protection of amino acids. It is also used to cleave such groups as Boc and Fmoc, and also to hydrogenolysis. It was used to prepare mononuclear nickel(II) acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Hydroxyethyl)pyridine is a metabolite of the anti-vertigo drug Betahistine. It acts as a reagent for protection of the 5′-phosphate of nucleotides as 2-pyridylethyl (Pet) esters. It is used for carboxyl protection of amino acids. It is also used to cleave such groups as Boc and Fmoc, and also to hydrogenolysis. It was used to prepare mononuclear nickel(II) acetate.

Solubility
Soluble in Chloroform (Slightly), Methanol (Slightly).

Notes
Hygroscopic. Store under dry inert gas. Store away from moisture, strong bases and oxidizing agents. Protect from humidity and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-74-2
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00006364
InChI Key BXGYBSJAZFGIPX-UHFFFAOYSA-N
Synonym 2-pyridineethanol, 2-2-hydroxyethyl pyridine, 2-2-pyridyl ethanol, pyridine-2-ethanol, 2-pyridin-2-yl ethan-1-ol, 2-pyridin-2-yl ethanol, 2-pyridinethanol, 2-pyridylethanol, 2-pyridine ethanol, 2-beta-hydroxyethyl pyridine
PubChem CID 7675
IUPAC Name 2-pyridin-2-ylethanol
SMILES C1=CC=NC(=C1)CCO

Specifications

Melting Point -8°C
Density 1.095
Boiling Point 235°C to 238°C
Flash Point 117°C (243°F)
Refractive Index 1.537
Quantity 100 g
Beilstein 111205
Sensitivity Hygroscopic
Solubility Information Soluble in Chloroform (Slightly),Methanol (Slightly).
Formula Weight 123.16
Percent Purity 99%
Chemical Name or Material 2-(2-Hydroxyethyl)pyridine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.