missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-N-(4-pyridinyl)propanamide, 97%, Thermo Scientific™

Product Code. p-2696645
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10364482

Brand: Thermo Scientific Maybridge MO00094DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 70298-89-4
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00996248
InChI Key JCMMVFHXRDNILC-UHFFFAOYSA-N
Synonym 2,2-dimethyl-n-pyridin-4-yl-propionamide, n-pyridin-4-yl pivalamide, 2,2-dimethyl-n-pyridin-4-yl propanamide, 2,2-dimethyl-n-4-pyridinyl propanamide, n-4-pyridyl pivalamide, 2,2-dimethyl-n-4-pyridyl propanamide, 4-2,2,2-trimethylacetamido pyridine, 2,2-dimethyl-n-4-pyridyl propionamide, 4-pivaloylaminopyridine, maybridge3_004407
PubChem CID 427059
IUPAC Name 2,2-dimethyl-N-pyridin-4-ylpropanamide
SMILES CC(C)(C)C(=O)NC1=CC=NC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material 2, 2-Dimethyl-N-(4-pyridinyl)propanamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.