missing translation for 'onlineSavingsMsg'
Learn More

2-[2-(Boc-amino)ethylamino]-3-iodopyridine, 95%, Alfa Aesar™

Product Code. 11434520
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 902835-89-6
Molecular Formula C12H18IN3O2
Molecular Weight (g/mol) 363.199
MDL Number MFCD07781137
InChI Key RHBATGHZRRPELS-UHFFFAOYSA-N
Synonym tert-butyl n-2-3-iodopyridin-2-yl amino ethyl carbamate, tert-butyl 2-3-iodopyridin-2-ylamino ethyl carbamate, 2-2-boc-amino ethylamino-3-iodopyridine, tert-butyl 2-3-iodopyridin-2-yl amino ethyl carbamate, tert-butoxy-n-2-3-iodo 2-pyridyl amino ethyl carboxamide
PubChem CID 28054477
IUPAC Name tert-butyl N-[2-[(3-iodopyridin-2-yl)amino]ethyl]carbamate
SMILES CC(C)(C)OC(=O)NCCNC1=C(C=CC=N1)I

Specifications

Quantity 250 mg
Formula Weight 363.2
Percent Purity 95%
Chemical Name or Material 2-[2-(Boc-amino)ethylamino]-3-iodopyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'