missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,4-Trimethylpentane, ≥99.5%, Honeywell™
Click to view available options
Quantity:
1 L
20 L
Packaging:
Glass Bottle
Specifications
Specifications
Density | 0.692g/mL at 25°C |
Quantity | 1 L |
Explosion Limit | 0.06 |
Ignition Point | 745°C |
Packaging | Glass Bottle |
Linear Formula | (CH3)2CHCH2C(CH3)3 |
Vapor Pressure | 41mmHg (21°C) |
UN Number | UN1262 |
Beilstein | 1696876 |
Formula Weight | 114.23g/mol |
Show More |
Safety and Handling
Product Identifier
- 2,2,4-Trimethylpentane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : 2,2,4-trimethylpentane
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'