Learn More
RETSCH 180° Stainless Steel Sieve For SR 200 Rotor Beater Mills
Shop All Retsch Products
Click to view available options
Pore Size:
0.08 mm
Unit Size:
Each
Specifications
Specifications
| Quantity | 1Each |
| Pore Size | 0.08 mm |
| Type | Sieve |
| Material | Stainless Steel |
| For Use With (Equipment) | SR 200 Rotor Beater Mills |