Learn More
Keystone Cleanroom Products 1422A Tyvek™ Equipment Cover
GREENER_CHOICE
1422A Tyvek™ Equipment Cover is an ideal choice in pharmaceutical operations to help keep clean equipment protected from both environmental and product cross contamination.
Brand:  Keystone Cleanroom Products COV-TVK-402448
 View more versions of this product
Product Code. 17758287
  • 5040.00€ / Pack of 50
Estimated Shipment: 22-04-2025
to see stock.
Explore more special offers
Description
  • Keeps cleaned equipment clean and protected from contamination
  • Breathable
  • Form fitting, fast and consistent
  • Manufactured and packaged in a ISO 6 cleanroom
  • Conveniently packaged: 1 heat-sealed labeled poly bag
  • Certificates of Conformance sent with shipment
Specifications
Specifications
Cover
Steam Sterilization
40 x 24 x 48 in.
101.6 cm
61 cm
121.9 cm
White
Tyvek
40 in.
24 in.
48 in.