Learn More
Keystone Cleanroom Products 1073B Tyvek™ Autoclaveable Wrapper Large
BIOPHARMAGREENER_CHOICE
Provides an excellent bacterial barrier for superior performance post steam sterilization. Cut using an automated CAD system in ISO 8 and made from DuPont 1073B Medical Grade Tyvek.
Brand:  Keystone Cleanroom Products MGTVKSHEET4040
 View more versions of this product
Product Code. 17728467
  • 779.00€ / Pack of 100
Estimated Shipment: 12-02-2025
to see stock.
Explore more special offers
Description
  • Designed for Steam Sterilization
  • Recommended for Use in Conjunction with a Secondary Tyvek™ Sterilization Pouch
  • Hydrophobic
  • Excellent Microbial Barrier
  • Resists Tearing and Punctures
  • Lint-Free - Category (Helmke Drum)
  • Manufactured and Packaged in a ISO 6 Cleanroom
  • Certificates of Conformance sent with shipment
Specifications
Specifications
Autoclavable
None
Single-use
Pharmaceutical Equipment
Non-sterile
<250°F
40 x 40 in.
101.6 cm
101.6 cm
COC
White
Steam Sterilization
Tyvek
Steam
<125°C
40 in.
40 in.