missing translation for 'onlineSavingsMsg'
Learn More

Boldenone Cypionate, Mikromol™

Product Code. 18082540
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 18082540

Brand: Mikromol MM3718.000100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Boldenone Cypionate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Boldenone Cypionate
CAS 106505-90-2
Analyte or Component Names Boldenone Cypionate
Shipping Condition Room Temperature
Molecular Formula C27 H38 O3
Certifications/Compliance ISO 17025
Synonym 17β-Hydroxyandrosta-1,4-dien-3-one Cyclopentanepropionate (7CI), Boldenone Cypionate
InChI Formula InChI=1S/C27H38O3/c1-26-15-13-20(28)17-19(26)8-9-21-22-10-11-24(27(22,2)16-14-23(21)26)30-25(29)12-7-18-5-3-4-6-18/h13,15,17-18,21-24H,3-12,14,16H2,1-2H3/t21-,22-,23-,24-,26-,27-/m0/s1
SMILES C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@@H]2OC(=O)CCC5CCCC5
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] 3-cyclopentylpropanoate
Molecular Weight (g/mol) 410.59
Quantity 100 mg
Formula Weight 410.2821 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.