missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-[4-(1,2-Diphenylethenyl)phenoxy]-N,N-diethylethanamine Hydrochloride, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
2-[4-(1,2-Diphenylethenyl)phenoxy]-N,N-diethylethanamine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | (E,Z)-2-[4-(1,2-diphenylethenyl)phenoxy]-N,N-diethylethanamine hydrochloride |
| CAS | 74056-26-1 |
| Analyte or Component Names | (E,Z)-2-[4-(1,2-diphenylethenyl)phenoxy]-N,N-diethylethanamine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H29 N O . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 2-[4-(1,2-Diphenylethenyl)phenoxy]-N,N-diethyl-ethanamine hydrochloride (1:1), Ethanamine, 2-[4-(1,2-diphenylethenyl)phenoxy]-N,N-diethyl-, hydrochloride (9CI), Triethylamine, 2-[p-(1,2-diphenylvinyl)phenoxy]-, hydrochloride, (E,Z)-2-[4-(1,2-diphenylethenyl)phenoxy]-N,N-diethylethanamine hydrochloride, 2-[4-(1,2-Diphenylethenyl)phenoxy]-N,N-diethylethanamine Hydrochloride, Clomiphene Related Compound A (USP), Clomiphene Related Compound A as Hydrochloride, Clomifene Citrate Impurity A as Hydrochloride, Clomifene Impurity A as Hydrochloride |
| InChI Formula | InChI=1S/C26H29NO.ClH/c1-3-27(4-2)19-20-28-25-17-15-24(16-18-25)26(23-13-9-6-10-14-23)21-22-11-7-5-8-12-22;/h5-18,21H,3-4,19-20H2,1-2H3;1H |
| SMILES | Cl.CCN(CC)CCOc1ccc(cc1)C(=Cc2ccccc2)c3ccccc3 |
| IUPAC Name | 2-[4-(1,2-diphenylethenyl)phenoxy]-N,N-diethylethanamine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction