missing translation for 'onlineSavingsMsg'
Learn More

10058-F4, Tocris Bioscience™

Product Code. 30001987
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Inhibits c-Myc-Max dimerization

10058-F4 is a cell permeable c-Myc-Max dimerization inhibitor. Inhibits proliferation, induces apoptosis and arrests cells in G0/G1 in rat1a-c-Myc cells. Also reduces tumor growth in vivo. In lymphoma cells, 10058-F4 decreases glucose uptake and expression of glycolysis-associated genes and reduces cell viability.

Chemical Identifiers

CAS 403811-55-2
Molecular Formula C12H11NOS2
Molecular Weight (g/mol) 249.346
InChI Key SVXDHPADAXBMFB-JXMROGBWSA-N
Synonym c-myc inhibitor, 5e-5-4-ethylbenzylidene-2-mercapto-1,3-thiazol-4 5h-one, 5-4-ethylphenyl methylene-2-thioxo-4-thiazolidinone, e-5-4-ethylbenzylidene-2-mercaptothiazol-4 5h-one, z,e-5-4-ethylbenzylidine-2-thioxothiazolidin-4-one, 5e-5-4-ethylphenyl methylidene-2-sulfanyl-1,3-thiazol-4-one, 5e-5-4-ethylphenyl methylidene-2-sulfanylidene-1,3-thiazolidin-4-one, 10058-f hplc , solid
PubChem CID 1271002
IUPAC Name (5E)-5-[(4-ethylphenyl)methylidene]-2-sulfanylidene-1,3-thiazolidin-4-one
SMILES CCC1=CC=C(C=C1)C=C2C(=O)NC(=S)S2

Specifications

Color Yellow
Quantity 10 mg
Formula Weight 249.35
Percent Purity >99%
Physical Form Solid
Chemical Name or Material 10058-F4
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'