Learn More
KGW Isotherm™ Dewar Cylindrical Insulated Flasks
For the storage and insulation of temperature-sensitive goods such as dry ice, liquid nitrogen and other coolants. KGW Isotherm™ Dewar Cylindrical Insulated Flasks feature a robust design made from borosilicate glass 3.3.
Brand:  KGW Isotherm™ 10211
 View more versions of this product
Product Code. 10433501
  • 215.00€ / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Easy handling
  • Cooling of small samples
  • Fully silvered as standard
  • Flexible applications in laboratory
  • Available with or without side grip
Specifications
Specifications
Blue
Cooling of small samples, storage of liquid nitrogen, dry ice, other coolants and as insulating flask
290mm
1 L
Borosilicate Glass
95mm
235mm
No side grip
77 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products