missing translation for 'onlineSavingsMsg'
Learn More

1-(Phenylsulfonyl)indole-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 15457217
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 278593-17-2
Molecular Formula C15H11NO4S
Molecular Weight (g/mol) 301.316
MDL Number MFCD02682021
InChI Key ISTBYLZYNVIQFS-UHFFFAOYSA-N
Synonym 1-phenylsulfonyl-1h-indole-3-carboxylic acid, 1-benzenesulfonyl indole-3-carboxylic acid, 1-phenylsulphonyl indole-3-carboxylic acid, 1-phenylsulfonyl indole-3-carboxylic acid, 1-benzenesulfonyl-1h-indole-3-carboxylic acid, 1h-indole-3-carboxylicacid, 1-phenylsulfonyl
PubChem CID 736484
IUPAC Name 1-(benzenesulfonyl)indole-3-carboxylic acid
SMILES C1=CC=C(C=C1)S(=O)(=O)N2C=C(C3=CC=CC=C32)C(=O)O

Specifications

Melting Point 236°C to 240°C
Quantity 1 g
Formula Weight 301.32
Percent Purity 97%
Chemical Name or Material 1-(Phenylsulfonyl)indole-3-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'