missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-n-octylimidazolium tetrafluoroborate, 99%, Thermo Scientific Chemicals

Product Code. 11416300
Click to view available options
Quantity:
5 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 244193-52-0
Molecular Formula C12H23BF4N2
Molecular Weight (g/mol) 282.13
MDL Number MFCD03095434
InChI Key GXZCAMSPWNHTAE-UHFFFAOYSA-N
Synonym 1-methyl-3-octylimidazolium tetrafluoroborate, 1-methyl-3-n-octylimidazolium tetrafluoroborate, 3-methyl-1-octylimidazolium tetrafluoroborate, 1-octyl-3-methylimidazolium tetrafluoroborate, omimbf4, dsstox_cid_27919, dsstox_rid_82672, dsstox_gsid_47943, 3-methyl-1-octylimidazoliumtetrafluoroborate, 1-methyl-3-octylimidazolium tetrafluoroborate hplc
PubChem CID 11460221
SMILES F[B-](F)(F)F.CCCCCCCCN1C=C[N+](C)=C1

Specifications

Density 1.12
Quantity 50 g
IUPAC Name 3-methyl-1-octyl-1H-imidazol-3-ium; tetrafluoroboranuide
Formula Weight 282.13
Percent Purity 99%
Chemical Name or Material 1-Methyl-3-n-octylimidazolium tetrafluoroborate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'