missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-methyl-1h-indazole-3-carbonyl chloride, 97%, Thermo Scientific™
90.20€ - 583.00€
Chemical Identifiers
CAS | 106649-02-9 |
---|---|
Molecular Formula | C9H7ClN2O |
Molecular Weight (g/mol) | 194.62 |
MDL Number | MFCD02093096 |
InChI Key | NPNWVMBPSINFBV-UHFFFAOYSA-N |
Synonym | 1-methyl-1h-indazole-3-carbonyl chloride, 1h-indazole-3-carbonylchloride, 1-methyl, 1-methyl-1h-indazol-3-carbonyl chloride, 1h-indazole-3-carbonyl chloride, 1-methyl, acmc-20mac8, 1-methyl-3-indazolylcarbonyl chloride, 1-methyl-indazole-3-carbonyl chloride, 1-methyl-1h-indazole-3-carboxychloride, 1-methyl-1h-indazole-3-carboxylic acid chloride |
PubChem CID | 7022055 |
IUPAC Name | 1-methylindazole-3-carbonyl chloride |
SMILES | CN1N=C(C(Cl)=O)C2=CC=CC=C12 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10015822
|
Thermo Scientific Maybridge
CC06302CB |
250 mg | Amber Glass Bottle |
90.20€
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10105512
|
Thermo Scientific Maybridge
CC06302DA |
1 g | Amber Glass Bottle |
257.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10215014
|
Thermo Scientific Maybridge
CC06302EA |
10 g | Amber Glass Bottle |
583.00€
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
106649-02-9 | |
194.62 | |
NPNWVMBPSINFBV-UHFFFAOYSA-N | |
7022055 | |
CN1N=C(C(Cl)=O)C2=CC=CC=C12 |
C9H7ClN2O | |
MFCD02093096 | |
1-methyl-1h-indazole-3-carbonyl chloride, 1h-indazole-3-carbonylchloride, 1-methyl, 1-methyl-1h-indazol-3-carbonyl chloride, 1h-indazole-3-carbonyl chloride, 1-methyl, acmc-20mac8, 1-methyl-3-indazolylcarbonyl chloride, 1-methyl-indazole-3-carbonyl chloride, 1-methyl-1h-indazole-3-carboxychloride, 1-methyl-1h-indazole-3-carboxylic acid chloride | |
1-methylindazole-3-carbonyl chloride |
Specifications
106649-02-9 | |
C9H7ClN2O | |
1-methyl-1h-indazole-3-carbonyl chloride, 1h-indazole-3-carbonylchloride, 1-methyl, 1-methyl-1h-indazol-3-carbonyl chloride, 1h-indazole-3-carbonyl chloride, 1-methyl, acmc-20mac8, 1-methyl-3-indazolylcarbonyl chloride, 1-methyl-indazole-3-carbonyl chloride, 1-methyl-1h-indazole-3-carboxychloride, 1-methyl-1h-indazole-3-carboxylic acid chloride | |
CN1N=C(C(Cl)=O)C2=CC=CC=C12 | |
194.62 | |
194.62 | |
1-Methyl-1H-indazole-3-carbonyl chloride |
97% | |
MFCD02093096 | |
NPNWVMBPSINFBV-UHFFFAOYSA-N | |
1-methylindazole-3-carbonyl chloride | |
7022055 | |
97% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title