missing translation for 'onlineSavingsMsg'
Learn More

1-Hexylamine, 99%

Product Code. p-7022757
Click to view available options
Quantity:
250 mL
1000 mL
Unit Size:
1000mL
250mL
This item is not returnable. View return policy

Product Code. 11467587

Brand: Thermo Scientific Alfa Aesar A15663.AK

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-26-2 | C6H15N | 101.19 g/mol

1-Hexylamine is used as corrosion inhibitors, emulsifiers and surfactants. It is used in dyes, rubber and pharmaceutical industries. It is also used to study the modification of alkanethlol monolayers in gas-phase surface reactions. It acts as an initiator in the ring opening polymerization of N-carboxyanhydride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Hexylamine is used as corrosion inhibitors, emulsifiers and surfactants. It is used in dyes, rubber and pharmaceutical industries. It is also used to study the modification of alkanethlol monolayers in gas-phase surface reactions. It acts as an initiator in the ring opening polymerization of N-carboxyanhydride.

Solubility
Miscible with water, dichloromethane, alcohol and acetone.

Notes
Air sensitive. Incompatible with acids, acid chlorides, carbon dioxide and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-26-2
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.19
MDL Number MFCD00008240
InChI Key BMVXCPBXGZKUPN-UHFFFAOYSA-N
Synonym hexylamine, 1-aminohexane, n-hexylamine, 1-hexanamine, 1-hexylamine, mono-n-hexylamine, hexanamine, hexyl amine, unii-ci4e002zv8, hexyl-amine
PubChem CID 8102
ChEBI CHEBI:5712
IUPAC Name hexan-1-amine
SMILES CCCCCCN

Specifications

Melting Point -19°C
Density 0.773
pH 11.6
Boiling Point 129°C to 131°C
Flash Point 27°C (80°F)
Odor Amine-like
Linear Formula CH3(CH2)5NH2
Refractive Index 1.418
Quantity 250 mL
UN Number UN2734
Beilstein 1731298
Sensitivity Air Sensitive
Solubility Information Miscible with water,dichloromethane,alcohol and acetone.
Formula Weight 101.19
Percent Purity 99%
Chemical Name or Material 1-Hexylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Hexylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : n-Hexylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.