missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Ethynylcyclohexylamine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
1-Ethynylcyclohexylamine was used in the synthesis of bicyclic and tetracyclic compounds. It was also used as a substrate in the reaction of a series of alkynes with mercury(II)acetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Ethynylcyclohexylamine was used in the synthesis of bicyclic and tetracyclic compounds. It was also used as a substrate in the reaction of a series of alkynes with mercury(II)acetate.
Notes
Air sensitive. Incompatible with oxidizing agents and acids.
1-Ethynylcyclohexylamine was used in the synthesis of bicyclic and tetracyclic compounds. It was also used as a substrate in the reaction of a series of alkynes with mercury(II)acetate.
Notes
Air sensitive. Incompatible with oxidizing agents and acids.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 0.913 |
Boiling Point | 65°C to 66°C (20 mmHg) |
Flash Point | 42°C (108°F) |
Refractive Index | 1.482 |
Quantity | 5 g |
UN Number | UN2734 |
Sensitivity | Air Sensitive |
Synonym | 1-ethynylcyclohexylamine, 1-ethynylcyclohexanamine, cyclohexanamine, 1-ethynyl, cyclohexanamine,1-ethynyl, 1-ethynyl-cyclohexylamine, 1-ethynylcyclohexaneamine, 1-ethynylcyclohexyl amine, 1-ethinyl-cyclohexyl-amine, 1-ethynyl-1-amino-cyclohexane |
Formula Weight | 123.2 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'