missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-3-methylimidazolium methyl sulfate, 98%, Thermo Scientific Chemicals

Product Code. 15425818
Click to view available options
Quantity:
5 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516474-01-4
Molecular Formula C7H14N2O4S
Molecular Weight (g/mol) 222.259
MDL Number MFCD08457697
InChI Key BXSDLSWVIAITRQ-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium methyl sulfate, 1-ethyl-3-methylimidazolium methylsulfate, 1-ethyl-3-methyl-1h-imidazol-3-ium methyl sulfate, 1-ethyl-3-methylimidazolium methyl sulfate 1-, 1-ethyl-3-methylimidazoliummethylsulfate, dsstox_cid_29266, dsstox_rid_83385, dsstox_gsid_49310, 1-ethyl-3-methylimidazolium methylsul-fate, 1-ethyl-3-methylimidazolium methyl sulfate hplc
PubChem CID 16211409
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;methyl sulfate
SMILES CCN1C=C[N+](=C1)C.COS(=O)(=O)[O-]

Specifications

Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 222.26
Percent Purity 98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium methyl sulfate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'