Learn More
Thermo Scientific Chemicals 1,6-Diiodoperfluorohexane, 97%
Brand:  Thermo Scientific Chemicals B21654.14
Additional Details : CAS : 375-80-4 Weight : 0.02500kg
 View more versions of this product
Product Code. 11309729
  • 447.00€ / 25g
In Stock
to see stock.
Description
Description
It is employed in the synthesis and characterization of fluorinated telomers containing vinylidene fluoride and hexafluoropropene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed in the synthesis and characterization of fluorinated telomers containing vinylidene fluoride and hexafluoropropene.
Notes
Light sensitive. Protect from light. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Specifications
1,6-Diiodoperfluorohexane
25°C
115°C (100 mmHg)
1.398
25 g
Light sensitive
JOQDDLBOAIKFQX-UHFFFAOYSA-N
1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodohexane
78994
97%
375-80-4
2.357
C6F12I2
MFCD00042264
1822186
1,6-diiodoperfluorohexane, 1,6-diiodododecafluorohexane, dodecafluoro-1,6-diiodohexane, 1,6-diiododecafluorohexane, perfluoro-1,6-diiodohexane, hexane, 1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodo, acmc-209iu0, 1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodo-hexane, hexane,1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro-1,6-diiodo
FC(F)(I)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
553.86
553.86
Product Suggestions
Product Suggestions
Videos
SDS Documents