missing translation for 'onlineSavingsMsg'
Learn More

1,4-Oxathiane, 98+%, Thermo Scientific Chemicals

Product Code. 11459196
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15980-15-1
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.167
MDL Number MFCD00006573
InChI Key JBYHSSAVUBIJMK-UHFFFAOYSA-N
Synonym 1,4-thioxane, p-thioxane, 1-oxa-4-thiacyclohexane, ccris 1746, 1,4-oxathiin, 2,3,5,6-tetrahydro, thioather, 1 pound not4-thioxane, 1-thia-4-oxacyclohexane, acmc-1bt7b, wln: t6o dstj
PubChem CID 27596
IUPAC Name 1,4-oxathiane
SMILES C1CSCCO1

Specifications

Density 1.114
Boiling Point 147°C to 148°C
Flash Point 42°C (107°F)
Odor Unpleasant
Refractive Index 1.507
Quantity 25 g
UN Number UN1993
Beilstein 102552
Formula Weight 104.17
Percent Purity ≥98%
Chemical Name or Material 1,4-Oxathiane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.