missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Diethylpiperazine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
1,4-Diethylpiperazine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4-Diethylpiperazine is used as pharmaceutical intermediate.
Solubility
Soluble in methanol, toluene.
Notes
store at a cool and dry place, with container tightly sealed. Store under nitrogen, recommended temperature is below 4°C.
1,4-Diethylpiperazine is used as pharmaceutical intermediate.
Solubility
Soluble in methanol, toluene.
Notes
store at a cool and dry place, with container tightly sealed. Store under nitrogen, recommended temperature is below 4°C.
Specifications
Specifications
Quantity | 10 g |
Synonym | piperazine, 1,4-diethyl, 1,4-diethyl piperazine, piperazine,1,4-diethyl, acmc-1be2o |
Solubility Information | Soluble in methanol,toluene. |
Formula Weight | 142.24 |
Percent Purity | 98% |
Chemical Name or Material | 1,4-Diethylpiperazine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'