missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Methoxyphenyl)piperazine, 95%, Thermo Scientific Chemicals

Product Code. 15411698
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16015-71-7
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
MDL Number MFCD00040733
InChI Key PZIBVWUXWNYTNL-UHFFFAOYSA-N
Synonym 1-3-methoxyphenyl piperazine, n-3-methoxyphenyl piperazine, 1-3-methoxyphenyl-piperazine, unii-6lcp52962g, 1-3-methoxy-phenyl-piperazine, chembl59597, piperazine, 1-3-methoxyphenyl, 3-methoxy-1-piperazinylbenzene, pubchem8585, 3-meopp
PubChem CID 81430
IUPAC Name 1-(3-methoxyphenyl)piperazine
SMILES COC1=CC=CC(=C1)N2CCNCC2

Specifications

Density 1.114
Boiling Point 150°C (0.5 mmHg)
Flash Point 91°C (196°F)
Refractive Index 1.5815
Quantity 5 g
UN Number UN3267
Formula Weight 192.26
Percent Purity 95%
Chemical Name or Material 1-(3-Methoxyphenyl)piperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'