missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester, 95%, Thermo Scientific Chemicals

Product Code. 15417655
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 903550-26-5
Molecular Formula C14H23BN2O3
Molecular Weight (g/mol) 278.159
MDL Number MFCD09037501
InChI Key ZZRFDLHBMBHJTI-UHFFFAOYSA-N
Synonym 1-tetrahydro-2h-pyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-2-tetrahydropyranyl-1h-pyrazole-5-boronic acid pinacol ester, 1-tetrahydro-2h-pyran-2-yl-1h-pyrazole-5-boronic acid pinacol ester, 1-tetrahydropyran-2-yl-1h-pyrazole-5-boronic acid pinacol ester, 1-oxan-2-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-oxan-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-oxan-2-yl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-oxan-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-y-1h-pyrazole, 1-tetrahydropyran-2-yl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole
PubChem CID 11587208
IUPAC Name 1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=NN2C3CCCCO3

Specifications

Quantity 250 mg
Formula Weight 278.16
Percent Purity 95%
Chemical Name or Material 1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.