missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Propanediol, 99+%, for analysis, Thermo Scientific Chemicals
46.43€ - 303.51€
Chemical Identifiers
CAS | 57-55-6 |
---|---|
Molecular Formula | C3H8O2 |
Molecular Weight (g/mol) | 76.09 |
MDL Number | MFCD00064272 |
InChI Key | DNIAPMSPPWPWGF-UHFFFAOYSA-N |
Synonym | 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost |
PubChem CID | 1030 |
ChEBI | CHEBI:16997 |
IUPAC Name | propane-1,2-diol |
SMILES | CC(CO)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10022910
|
Thermo Scientific Acros
220870250 |
25 mL | Glass bottle |
46.43€
25mL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
10553114
|
Thermo Scientific Acros
220872500 |
250 mL | Glass bottle |
54.07€
250mL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
10154510
|
Thermo Scientific Acros
220870010 |
1 L | Glass bottle |
159.80€
1 L |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
10307243
|
Thermo Scientific Acros
220870025 |
2.5 L | Glass bottle |
303.51€
2.50 L |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
57-55-6 | |
76.09 | |
DNIAPMSPPWPWGF-UHFFFAOYSA-N | |
1030 | |
propane-1,2-diol |
C3H8O2 | |
MFCD00064272 | |
1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost | |
CHEBI:16997 | |
CC(CO)O |
Specifications
57-55-6 | |
100.0 | |
1.0300g/mL | |
99°C | |
99% min. (GC) | |
CH3CH(OH)CH2OH | |
MFCD00064272 | |
15, 7968 | |
1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost | |
DNIAPMSPPWPWGF-UHFFFAOYSA-N | |
propane-1,2-diol | |
1030 | |
45 mPa.s (20°C) | |
99+% | |
Viscous Liquid |
99.0 | |
-60.0°C | |
187.0°C | |
Authentic | |
C3H8O2 | |
1.4310 to 1.4330 | |
01, 472 | |
1.03 | |
Solubility in water: completely soluble. Other solubilities: soluble in ether, miscible with acetone and chloroform | |
CC(CO)O | |
76.09 | |
CHEBI:16997 | |
76.09 | |
Analysis | |
1, 2-Propanediol |
Safety and Handling
missing translation for 'einecsNumber' : 200-338-
RUO – Research Use Only