missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Bis[(2S,5S)-2,5-diphenyl-1-phospholanyl]ethane, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
It is a DuPhos and BPE ligands which are highly efficient privileged ligands. It is a chiral phosphine ligand used in the asymmetric hydrogenation of allylic sulfoxides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is a DuPhos and BPE ligands which are highly efficient privileged ligands. It is a chiral phosphine ligand used in the asymmetric hydrogenation of allylic sulfoxides.
Solubility
Sparingly Soluble (3.2E-5 g/L) (25°C).
Notes
Incompatible with strong oxidizing agents. Store under room temperature. Air sensitive.
It is a DuPhos and BPE ligands which are highly efficient privileged ligands. It is a chiral phosphine ligand used in the asymmetric hydrogenation of allylic sulfoxides.
Solubility
Sparingly Soluble (3.2E-5 g/L) (25°C).
Notes
Incompatible with strong oxidizing agents. Store under room temperature. Air sensitive.
Specifications
Specifications
Melting Point | 143°C to 147°C |
Quantity | 250 mg |
Sensitivity | Air Sensitive |
Solubility Information | Sparingly Soluble (3.2E-5 g/L) (25°C). |
Formula Weight | 506.6 |
Percent Purity | ≥97% |
Chemical Name or Material | 1,2-Bis[(2S,5S)-2,5-diphenyl-1-phospholanyl]ethane |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'