ULT Freezers - Request a Call Back
Do you need an Ultra Low Temperature Freezer for storage of vaccines or another application?
Speak to a specialist to receive more information about products and services for cold storage equipment.
To see our full list of ULT Freezers, please see below.
BINDER™ Series UF V - Ultra-Low Temperature Freezer, with Climate-Neutral Refrigerants
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice Product
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
The Series UF V - Ultra-Low Temperature Freezer ensures the safe storage of samples at -90°C. It combines outstanding environmental friendliness with low energy consumption, convenient operation and an individual safety concept.
Promotions are available
Insulation | The combination of vacuum insulation panels (V technology) and CFC-free polyurethane foaming |
---|---|
Plug Type | NEMA 5-15 P |
Door Style | Foamed |
Refrigerant | Stage 1: R290, Stage 2: R170 |
Chamber Material | Stainless Steel |
Wattage | 1.6 kW |
Cabinet Material | Anti-corrosion Steel |
Compressor | Bottom-mounted |
Monitoring Options | Integrated Alarm Systems |
Type | Ultra Low temperature Freezer |
Voltage | 230 V |
Shelves | 3 |
Frequency | 50 Hz |
Temperature Range | -40°C to -86°C |
No. of Doors | 1 |
Holds | 12 |
Display | LCD |
Fisherbrand™ Isotemp™ Ultra Low Temperature Freezer
Fisherbrand™ Isotemp™ Ultra Low Temperature Freezer is dependable for storage applications with energy and space savings features.
Promotions are availableInsulation | Vacuum, Foam |
---|---|
Warranty | US: 2 years parts and labor + additional 2 years compressor parts CA: 5 year standard warranty |
Refrigerant | R290 / R170+R290 Mix |
Height (English) Exterior | 77.99 in. |
Color | Gray |
Cabinet Material | Cold-rolled Steel |
Interior | Painted Steel |
Breaker | Yes |
For Use With (Application) | Long term storage |
Typical Lifetime | 10 yrs |
Height (Metric) Exterior | 198.1 cm |
Condenser Type | Air Cooled |
Temperature Sensor | RTD Probe |
Type | Ultra-Low Freezer, Upright |
Setpoint Security | Yes |
Casters | 4 |
Defrost | Manual |
Style | Upright |
Temperature Range | -50°C to -86°C |
Exterior Finish | Painted Steel |
Display | Capacitive Touch |
Stirling Ultracold Portable/Deployable Ultra-Low Temperature Freezer
The ULT25NEU is a portable ULT freezer solution for bringing ULT storage to the patient. This small, lightweight, and efficient portable -86°C freezer is ideal for clinical trials and biologic drug delivery.
Eppendorf™ CryoCube™ F740
Greener Choice Product
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
Polyurethane foam insulation combined with a highly efficient cooling system achieve and maintain ultra-low temperatures. Eppendorf™ CryoCube™ F740 is known for its energy efficiency, reliability, and worry-free, long-term sample preservation.
Insulation | Vacuum, Polyurethane Foam |
---|---|
Electrical Requirements | 230 V 50 Hz |
Warranty | 5 years |
Capacity (Metric) | 740 L |
Certifications/Compliance | CE |
Inner Doors | 3 |
Eppendorf™ CryoCube F740hiw ULT Freezer, 3 Compartments, CO2 Backup
Greener Choice Product
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
Innovative, highest quality, minus 86 °C ultra-low temperature freezer featuring green cooling, reduced power consumption, and a variety of configurable options.
Electrical Requirements | Integrated voltage inverter for flexible usage with 100 – 230 V mains / power supply. |
---|---|
Description | Water-Cooled |
Door Style | Left Handle |
Refrigerant | R290/R170: Environmentally safe, HCFC-free and CFC-free refrigerants |
Dimensions (D x W x H) Interior | 62.1 x 86.5 x 139 cm |
Capacity (Metric) | 740 L |
Backup Systems | CO2 (factory installed) |
Cabinet Material | Stainless Steel |
Array | 3 Inner Compartments |
Water Cooled | Yes |
Casters | Yes |
Model | Left-Handled Door Model |
Chart Recorder | No |
Certifications/Compliance | Individual Certificate of Quality, complete with serial number, provided as standard for your documentation. |
Dimensions (DxWxH) Exterior | 98 x 109.9 x 197.3 cm |
Eppendorf™ CryoCube F740hi ULT Freezer, 5 Compartments, LN2 Backup
Greener Choice Product
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
Innovative, highest quality, minus 86 °C ultra-low temperature freezer featuring green cooling, reduced power consumption, and a variety of configurable options.
Array | 5 Inner Compartments |
---|---|
Water Cooled | No |
Electrical Requirements | Integrated voltage inverter for flexible usage with 100 – 230 V mains / power supply. |
Description | Air-Cooled |
Casters | Yes |
Refrigerant | R290/R170: Environmentally safe, HCFC-free and CFC-free refrigerants |
Dimensions (D x W x H) Interior | 62.1 x 86.5 x 139 cm |
Capacity (Metric) | 740 L |
Backup Systems | LN2 (factory installed) |
Cabinet Material | Stainless Steel |
Chart Recorder | No |
Certifications/Compliance | Individual Certificate of Quality, complete with serial number, provided as standard for your documentation. |
Dimensions (DxWxH) Exterior | 98 x 109.9 x 197.3 cm |
Eppendorf™ CryoCube F740hiw ULT Freezer, 5 Compartments, LN2 Backup
Greener Choice Product
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
Innovative, highest quality, minus 86 °C ultra-low temperature freezer featuring green cooling, reduced power consumption, and a variety of configurable options.
Electrical Requirements | Integrated voltage inverter for flexible usage with 100 – 230 V mains / power supply. |
---|---|
Description | Water-Cooled |
Door Style | Left Handle |
Refrigerant | R290/R170: Environmentally safe, HCFC-free and CFC-free refrigerants |
Dimensions (D x W x H) Interior | 62.1 x 86.5 x 139 cm |
Capacity (Metric) | 740 L |
Backup Systems | LN2 (factory installed) |
Cabinet Material | Stainless Steel |
Array | 5 Inner Compartments |
Water Cooled | Yes |
Casters | Yes |
Model | Left-Handled Door Model |
Chart Recorder | No |
Certifications/Compliance | Individual Certificate of Quality, complete with serial number, provided as standard for your documentation. |
Dimensions (DxWxH) Exterior | 98 x 109.9 x 197.3 cm |
Eppendorf™ CryoCube F740hi ULT Freezer, 5 Compartments, CO2 Backup
Greener Choice Product
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
Innovative, highest quality, minus 86 °C ultra-low temperature freezer featuring green cooling, reduced power consumption, and a variety of configurable options.
Array | 5 Inner Compartments |
---|---|
Water Cooled | No |
Electrical Requirements | Integrated voltage inverter for flexible usage with 100 – 230 V mains / power supply. |
Description | Air-Cooled |
Casters | Yes |
Refrigerant | R290/R170: Environmentally safe, HCFC-free and CFC-free refrigerants |
Dimensions (D x W x H) Interior | 62.1 x 86.5 x 139 cm |
Capacity (Metric) | 740 L |
Backup Systems | CO2 (factory installed) |
Cabinet Material | Stainless Steel |
Chart Recorder | No |
Certifications/Compliance | Individual Certificate of Quality, complete with serial number, provided as standard for your documentation. |
Dimensions (DxWxH) Exterior | 98 x 109.9 x 197.3 cm |
Eppendorf™ CryoCube F740hi ULT Freezer, 3 Compartments, CO2 Backup
Greener Choice Product
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
Innovative, highest quality, minus 86 °C ultra-low temperature freezer featuring green cooling, reduced power consumption, and a variety of configurable options.
Array | 3 Inner Compartments |
---|---|
Water Cooled | No |
Electrical Requirements | Integrated voltage inverter for flexible usage with 100 – 230 V mains / power supply. |
Description | Air-Cooled |
Casters | Yes |
Refrigerant | R290/R170: Environmentally safe, HCFC-free and CFC-free refrigerants |
Dimensions (D x W x H) Interior | 62.1 x 86.5 x 139 cm |
Capacity (Metric) | 740 L |
Backup Systems | CO2 (factory installed) |
Cabinet Material | Stainless Steel |
Chart Recorder | No |
Certifications/Compliance | Individual Certificate of Quality, complete with serial number, provided as standard for your documentation. |
Dimensions (DxWxH) Exterior | 98 x 109.9 x 197.3 cm |
CryoCube FC660h Chest Freezer
Greener Choice Product
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
Use this ultra-low temperature freezer for storage and preservation of all types of biological samples. New Brunswick Scientific™ CryoCube FC660h Chest Freezers feature a high efficiency fan, compressor and condenser which decrease power consumption.
Eppendorf™ CryoCube F740hiw ULT Freezer, 3 Compartments, LN2 Backup
Greener Choice Product
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
Innovative, highest quality, minus 86 °C ultra-low temperature freezer featuring green cooling, reduced power consumption, and a variety of configurable options.
Electrical Requirements | Integrated voltage inverter for flexible usage with 100 – 230 V mains / power supply. |
---|---|
Description | Water-Cooled |
Door Style | Left Handle |
Refrigerant | R290/R170: Environmentally safe, HCFC-free and CFC-free refrigerants |
Dimensions (D x W x H) Interior | 62.1 x 86.5 x 139 cm |
Capacity (Metric) | 740 L |
Backup Systems | LN2 (factory installed) |
Cabinet Material | Stainless Steel |
Array | 3 Inner Compartments |
Water Cooled | Yes |
Casters | Yes |
Model | Left-Handled Door Model |
Chart Recorder | No |
Certifications/Compliance | Individual Certificate of Quality, complete with serial number, provided as standard for your documentation. |
Dimensions (DxWxH) Exterior | 98 x 109.9 x 197.3 cm |
Eppendorf™ CryoCube F740hi ULT Freezer, 3 Compartments, LN2 Backup
Greener Choice Product
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
Innovative, highest quality, minus 86 °C ultra-low temperature freezer featuring green cooling, reduced power consumption, and a variety of configurable options.
Array | 3 Inner Compartments |
---|---|
Water Cooled | No |
Electrical Requirements | Integrated voltage inverter for flexible usage with 100 – 230 V mains / power supply. |
Description | Air-Cooled |
Casters | Yes |
Refrigerant | R290/R170: Environmentally safe, HCFC-free and CFC-free refrigerants |
Dimensions (D x W x H) Interior | 62.1 x 86.5 x 139 cm |
Capacity (Metric) | 740 L |
Backup Systems | LN2 (factory installed) |
Cabinet Material | Stainless Steel |
Chart Recorder | No |
Certifications/Compliance | Individual Certificate of Quality, complete with serial number, provided as standard for your documentation. |
Dimensions (DxWxH) Exterior | 98 x 109.9 x 197.3 cm |
Eppendorf™ CryoCube F740hiw ULT Freezer, 5 Compartments, CO2 Backup
Greener Choice Product
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
Innovative, highest quality, minus 86 °C ultra-low temperature freezer featuring green cooling, reduced power consumption, and a variety of configurable options.
Electrical Requirements | Integrated voltage inverter for flexible usage with 100 – 230 V mains / power supply. |
---|---|
Description | Water-Cooled |
Door Style | Left Handle |
Refrigerant | R290/R170: Environmentally safe, HCFC-free and CFC-free refrigerants |
Dimensions (D x W x H) Interior | 62.1 x 86.5 x 139 cm |
Capacity (Metric) | 740 L |
Backup Systems | CO2 (factory installed) |
Cabinet Material | Stainless Steel |
Array | 5 Inner Compartments |
Water Cooled | Yes |
Casters | Yes |
Model | Left-Handled Door Model |
Chart Recorder | No |
Certifications/Compliance | Individual Certificate of Quality, complete with serial number, provided as standard for your documentation. |
Dimensions (DxWxH) Exterior | 98 x 109.9 x 197.3 cm |
Eppendorf™ Innova Upright Freezers
Greener Choice Product
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
Use this freezer for storage and preservation of all types of biological material. New Brunswick Scientific™ Innova upright Freezers combine classical insulation foam with vacuum insulation panels to offer long lasting and reliable ULT technology for safe sample storage.
Thermo Scientific™ TSX Series Ultra-Low Freezers
SureTRACE
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
Greener Choice Product
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
Supports traceability with guaranteed access to certificates and proactive change notifications.
Learn More
This product offers one or more environmental benefits itemized in the U.S. FTC “Green Guides.”
Learn More About the Greener Choice Program
Thermo Scientific™ TSX Series ultra-low freezers are designed to meet the highest sustainability, temperature management and reliability standards for -80°C and -70°C storage.
Insulation | Vacuum Insulation Panels (VIP) + Water-Blown Polyurethane Foam (SNAP-compliant) |
---|---|
Warranty | Warranty varies by country, please contact us for details |
Height (Metric) Interior | 130.1 cm |
Smart View Compatibility | Smart-Vue Compatible |
Refrigerant | R290 (1st stage), R170 + R290 mix (2nd stage) |
Height (English) Exterior | 78 in. |
Compressor | V-drive |
Interior | Stainless Steel |
Monitoring Options | Alarm, Recorder |
Height (Metric) Exterior | 198.1 cm |
Depth (English) Exterior | 37.6 in. |
Depth (Metric) Interior | 71.9 cm |
Type | Ultra-Low Freezer, Upright |
Defrost | Manual |
Shelves | 3 |
Connectivity | Thermo Scientific™ DeviceLink™ (Networks: WPA2-PSK, WPA-PSK, WEP, WPA2-EAP-PEAP, WPA-EAP-PEAP) |
Style | Upright |
Temperature Range | -50°C to -86°C |
Depth (Metric) Exterior | 95.5 cm |
Depth (English) Interior | 28.3 in. |
Height (English) Interior | 51.2 in. |
Product Line | TSX Series |
Display | Touch-Screen LCD |