accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 78-83-1

CH3H3CHO

CAS RN 78-83-1

IUPAC Name: 2-methylpropan-1-ol
Synonyms: isobutanol titanium (+4) salt isobutyl alcohol fema number 2179 2-methylpropanoi fermentation butyl alcohol IBA isobutanol or isobutyl alcohol isobutylalkohol aluminum salt isobutyl alcohol I-butyl alcohol
Molecular Weight (g/mol): 74.12
Molecular Formula: C4H10O
InChi Key: ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
SMILES: CC(C)CO
Quantity 
  • (11)
  • (1)
  • (7)
  • (3)
  • (1)
  • (2)
  • (3)
  • (6)
Boiling Point 
  • (11)
  • (13)
  • (8)
Molecular Weight (g/mol) 
  • (13)
  • (19)
Grade 
  • (3)
  • (5)
  • (2)
  • (2)
  • (3)
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (10)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (5)
  • (1)
  • (6)
  • (1)
  • (1)
  • (10)
  • (11)
  • (11)
  • (1)
  • (7)
  • (3)
  • (1)
  • (2)
  • (3)
  • (6)
  • (11)
  • (13)
  • (8)
  • (13)
  • (19)
  • (3)
  • (5)
  • (2)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (10)
  • (17)
  • (1)
  • (1)
  • (1)