accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6004-24-6

H2OClH3CNCH

CAS RN 6004-24-6

IUPAC Name: 1-hexadecyl-1,4-dihydropyridin-4-ylium hydrate chloride
Synonyms: 1-hexadecyl-4H-pyridin-4-ylium hydrate chloride
Molecular Weight (g/mol): 358.01
Molecular Formula: C21H40ClNO
InChi Key: NFCRBQADEGXVDL-UHFFFAOYSA-M
SMILES: O.[Cl-].CCCCCCCCCCCCCCCCN1C=C[CH+]C=C1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)