accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 580-19-8

H2NN

CAS RN 580-19-8

IUPAC Name: quinolin-7-amine
Synonyms: quinolin-7-amine
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: RZAUIOKDXQWSQE-UHFFFAOYSA-N
SMILES: NC1=CC=C2C=CC=NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results