accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5459-40-5

CH3OCH2

CAS RN 5459-40-5

IUPAC Name: 1-ethenyl-4-ethoxybenzene
Synonyms: 1-ethenyl-4-ethoxybenzene
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: OBRYRJYZWVLVLF-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results