man

Test

CAS RN 203310-42-3

CH3SOON

CAS RN 203310-42-3

IUPAC Name: 2-(hexane-1-sulfonyl)acetonitrile
Synonyms: 2-(hexane-1-sulfonyl)acetonitrile
Molecular Weight (g/mol): 189.27
Molecular Formula: C8H15NO2S
InChi Key: CGKSNJYMJMDPQT-UHFFFAOYSA-N
SMILES: CCCCCCS(=O)(=O)CC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results