man

Test

CAS RN 1519-47-7

ClClPP

CAS RN 1519-47-7

IUPAC Name: triphenyl({4-[(triphenylphosphaniumyl)methyl]phenyl}methyl)phosphanium dichloride
Synonyms: triphenyl({4-[(triphenylphosphaniumyl)methyl]phenyl}methyl)phosphanium dichloride
Molecular Weight (g/mol): 699.64
Molecular Formula: C44H38Cl2P2
InChi Key: IUNJPQCPINLADS-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].C(C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results