accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 143-28-2

H3COH(Z)

CAS RN 143-28-2

IUPAC Name: (9Z)-octadec-9-en-1-ol
Synonyms: conditioner 1 cachalot O-15 ocenol HD oleyl alcohol 80/85 dermaffine loxanol 95 oleo alcohol siponol OC loxanol M cachalot O-8
Molecular Weight (g/mol): 268.49
Molecular Formula: C18H36O
InChi Key: ALSTYHKOOCGGFT-KTKRTIGZSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCCO
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (5)