man

Test

CAS RN 104376-79-6

H2OH2OH2OH2OH2OH2OH2ONaNaNaNaH3CONOHNSNOOOSNOONNH3CSNH2NH3CONOHNSNOOOSNONNH3COSNH2N

CAS RN 104376-79-6

IUPAC Name: tetrasodium 3-({[(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-2-carboxylato-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}sulfanyl)-2-methyl-5,6-dioxo-1,2,5,6-tetrahydro-1,2,4-triazin-1-ide (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-{[(2-methyl-6-oxido-5-oxo-2,5-dihydro-1,2,4-triazin-3-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate heptahydrate
Synonyms: tetrasodium 3-({[(6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-2-carboxylato-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-en-3-yl]methyl}sulfanyl)-2-methyl-5,6-dioxo-1H-1,2,4-triazin-1-ide (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-{[(2-methyl-6-oxido-5-oxo-1,2,4-triazin-3-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate heptahydrate
Molecular Weight (g/mol): 1323.17
Molecular Formula: C36H46N16Na4O21S6
InChi Key: RGDBKADCOSIOEV-MAODNAKNSA-J
SMILES: O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC3=NC(=O)C(=O)[N-]N3C)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1.CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC3=NC(=O)C([O-])=NN3C)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
Type 
  • (3)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)